2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole 97%

Supplier: Apollo Scientific
Warning

PC5065-1G PC5065-500MG PC5065-5G
APOSPC5065-1GEA 72.5 EUR
APOSPC5065-1G APOSPC5065-500MG APOSPC5065-5G
2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole 97%
2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole
Formula: C₄H₄F₃N₃S
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00042321
CAS Number: 25366-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR