4,4,4-Trifluoro-3-methyl-3-trimethylsilyloxybutan-2-one

Supplier: Apollo Scientific

PC50974-1G PC50974-250MG
APOSPC50974-1GEA 522 EUR
APOSPC50974-1G APOSPC50974-250MG
4,4,4-Trifluoro-3-methyl-3-trimethylsilyloxybutan-2-one
4,4,4-Trifluoro-3-methyl-3-trimethylsilyloxybutan-2-one
Formula: C₈H₁₅F₃O₂Si
MW: 228.284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32693997
CAS Number: 2649323-00-0
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR