2-(5,7-Difluoro-6-quinolyl)propanoic acid

Supplier: Apollo Scientific

PC510018-1G
APOSPC510018-1GEA 601 EUR
APOSPC510018-1G
2-(5,7-Difluoro-6-quinolyl)propanoic acid
2-(5,7-Difluoro-6-quinolyl)propanoic acid
Formula: C₁₂H₉F₂NO₂
MW: 237,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16495907
CAS Number: 1226776-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR