1-(2-Fluorophenyl)-4-oxocyclohexanecarboxylic acid

Supplier: Apollo Scientific

PC510131-5G
APOSPC510131-5GEA 1010 EUR
APOSPC510131-5G
1-(2-Fluorophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Fluorophenyl)-4-oxocyclohexanecarboxylic acid
Formula: C₁₃H₁₃FO₃
MW: 236.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17013935
CAS Number: 179064-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR