Ethyl 2,2-Difluoro-3-(methylamino)propanoate hydrochloride

Supplier: Apollo Scientific

PC510175-1G
APOSPC510175-1GEA 804 EUR
APOSPC510175-1G
Ethyl 2,2-Difluoro-3-(methylamino)propanoate hydrochloride
Ethyl 2,2-Difluoro-3-(methylamino)propanoate hydrochloride
Formula: C₆H₁₁F₂NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD27986811
CAS Number: 1956311-12-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR