4-Fluoro-4-(m-tolyl)piperidine

Supplier: Apollo Scientific

PC510219-1G
APOSPC510219-1GEA 1010 EUR
APOSPC510219-1G
4-Fluoro-4-(m-tolyl)piperidine
4-Fluoro-4-(m-tolyl)piperidine
Formula: C₁₂H₁₆FN
MW: 193,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26037730
CAS Number: 1547092-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR