6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole

Supplier: Apollo Scientific
Warning

PC51022-1G PC51022-250MG
APOSPC51022-1GEA 609 EUR
APOSPC51022-1G APOSPC51022-250MG
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole
Formula: C₈H₅BrFIN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18384874
CAS Number: 1286734-96-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR