2-(4-Fluorophenyl)-2-hydroxypropionic acid

Supplier: Apollo Scientific

PC510273-1G
APOSPC510273-1GEA 734 EUR
APOSPC510273-1G
2-(4-Fluorophenyl)-2-hydroxypropionic acid
2-(4-Fluorophenyl)-2-hydroxypropionic acid
Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD11052520
CAS Number: 81170-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR