3-Amino-5-fluoro-1H-indazole

Supplier: Apollo Scientific
Warning

PC51032-1G PC51032-250MG
APOSPC51032-1GEA 386 EUR
APOSPC51032-1G APOSPC51032-250MG
3-Amino-5-fluoro-1H-indazole
3-Amino-5-fluoro-1H-indazole
Formula: C₇H₆FN₃
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD03426699
CAS Number: 61272-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR