Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate

Supplier: Apollo Scientific

PC510333-5G
APOSPC510333-5GEA 1010 EUR
APOSPC510333-5G
Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate
Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate
Formula: C₁₃H₁₄F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD29065894
CAS Number: 924312-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR