2-Fluoro-5-hydroxy-3-methylpyridine 97%

Supplier: Apollo Scientific
Warning

PC51123-250MG
APOSPC51123-250MGEA 21.9 EUR
APOSPC51123-250MG
2-Fluoro-5-hydroxy-3-methylpyridine 97%
2-Fluoro-5-hydroxy-3-methylpyridine
Formula: C₆H₆FNO
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD09839279
CAS Number: 186593-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR