2-Fluoro-3-hydroxy-5-methylpyridine

Supplier: Apollo Scientific
Warning

PC51125-500MG
APOSPC51125-500MGEA 157 EUR
APOSPC51125-500MG
2-Fluoro-3-hydroxy-5-methylpyridine
2-Fluoro-3-hydroxy-5-methylpyridine
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD16611070
CAS Number: 1184172-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR