Methyl 2-(difluoromethyl)isonicotinate

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Difluoromethyl)-4-(methoxycarbonyl)pyridine, Methyl 2-(difluoromethyl)pyridine-4-carboxylate

PC51205-1G PC51205-250MG
APOSPC51205-1GEA 1400 EUR
APOSPC51205-1G APOSPC51205-250MG
Methyl 2-(difluoromethyl)isonicotinate
Methyl 2-(difluoromethyl)isonicotinate
Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22690248
CAS Number: 1251844-44-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR