(5-Amino-2-fluorophenyl)methanol 99%

Supplier: Apollo Scientific
Warning

PC5127-1G PC5127-5G PC5127-25G
APOSPC5127-1GEA 21.9 EUR
APOSPC5127-1G APOSPC5127-5G APOSPC5127-25G
(5-Amino-2-fluorophenyl)methanol 99%
(5-Amino-2-fluorophenyl)methanol
Formula: C₇H₈FNO
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD03094257
CAS Number: 84832-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR