Methyl trifluoromethanesulphonate 98%

Supplier: Apollo Scientific
Danger

PC5170-10G PC5170-50G PC5170-100G
APOSPC5170-10GEA 29 EUR
APOSPC5170-10G APOSPC5170-50G APOSPC5170-100G
Methyl trifluoromethanesulphonate 98%
Methyl trifluoromethanesulphonate
Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 94…102 °C
Melting Pt: –64 °C
Density: 1.45 g/cm³ (25 °C)
Flash Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000409
CAS Number: 333-27-7
EINECS: 206-371-7
UN: 2920
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR