Methyl-3-(trifluoromethyl)benzoate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3-(trifluoromethyl)benzoate

PC5171Z-100G PC5171Z-25G PC5171Z-500G
APOSPC5171Z-100GEA 46.4 EUR
APOSPC5171Z-100G APOSPC5171Z-25G APOSPC5171Z-500G
Methyl-3-(trifluoromethyl)benzoate 95%
Methyl 3-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 93…95 °C/20mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR