5-(Difluoromethyl)-2-methyl-pyrimidin-4-amine

Supplier: Apollo Scientific

PC520056-1G
APOSPC520056-1GEA 3190 EUR
APOSPC520056-1G
5-(Difluoromethyl)-2-methyl-pyrimidin-4-amine
5-(Difluoromethyl)-2-methyl-pyrimidin-4-amine
Formula: C₆H₇F₂N₃
Storage Temperature: Ambient
MDL Number: MFCD29767539
CAS Number: 1785079-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR