N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%

Supplier: Apollo Scientific
Danger

PC520402-100MG PC520402-1G PC520402-250MG
APOSPC520402-100MGEA 21.9 EUR
APOSPC520402-100MG APOSPC520402-1G APOSPC520402-250MG
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula: C₁₁H₁₂FN·ClH
Storage Temperature: Freezer
MDL Number: MFCD31745724
CAS Number: 2379918-37-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR