6-Ethoxy-2,3-difluoroaniline 95%

Supplier: Apollo Scientific

PC52456-1G PC52456-250MG PC52456-5G
APOSPC52456-1GEA 497 EUR
APOSPC52456-1G APOSPC52456-250MG APOSPC52456-5G
6-Ethoxy-2,3-difluoroaniline 95%
6-Ethoxy-2,3-difluoroaniline
Formula: C₈H₉F₂NO
Storage Temperature: Ambient
CAS Number: 2385070-70-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR