2,6-Dichloro-4-(trifluoromethoxy)phenylacetonitrile

Supplier: Apollo Scientific

PC52462-1G PC52462-5G
APOSPC52462-1GEA 190 EUR
APOSPC52462-1G APOSPC52462-5G
2,6-Dichloro-4-(trifluoromethoxy)phenylacetonitrile
2,6-Dichloro-4-(trifluoromethoxy)phenylacetonitrile
Formula: C₉H₄Cl₂F₃NO
Storage Temperature: Ambient
CAS Number: 886503-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR