α,α,α-Trifluoro-3-nitrotoluene 95%

Supplier: Apollo Scientific
Danger

PC5250-100G PC5250-1KG PC5250-500G PC5250-25G PC5250-5KG PC5250-10KG
APOSPC5250-100GEA 69.6 EUR
APOSPC5250-100G APOSPC5250-1KG APOSPC5250-500G APOSPC5250-25G APOSPC5250-5KG APOSPC5250-10KG
α,α,α-Trifluoro-3-nitrotoluene 95%
α,α,α-Trifluoro-3-Nitrotoluene
Formula: C₇H₄F₃NO₂
Boiling Pt: 200…205 °C
Melting Pt: –5 °C
Density: 1,436 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007260
CAS Number: 98-46-4
EINECS: 202-670-1
UN: 2306
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR