Nitrosium tetrafluoroborate 98%

Supplier: Apollo Scientific
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

PC5290-100G
APOSPC5290-100GEA 654 EUR
APOSPC5290-100G
Nitrosium tetrafluoroborate 98%
Nitrosium tetrafluoroborate

Versatile crystalline nitrosating agent. Converts arylamines to aryl fluorides via diazonium tetrafluoroborates.

Formula: BF₄NO
MW: 116.81 g/mol
Melting Pt: 250 °C (subl.)
Density: 2.185 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR