4-Trifluoromethylphenethyl bromide 97%

Supplier: Apollo Scientific

Synonyms: 1-(2-Bromoethyl)-4-(trifluoromethyl)benzene

PC530024-1G PC530024-5G
APOSPC530024-1GEA 77 EUR
APOSPC530024-1G APOSPC530024-5G
4-Trifluoromethylphenethyl bromide 97%
4-Trifluoromethylphenethyl bromide
Formula: C₉H₈BrF₃
Density: 1,499 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD10567022
CAS Number: 130365-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR