(S)-b-Amino-3-fluorobenzenepropanol 97%

Supplier: Apollo Scientific

PC530051-1G PC530051-250MG
APOSPC530051-1GEA 579 EUR
APOSPC530051-1G APOSPC530051-250MG
(S)-b-Amino-3-fluorobenzenepropanol 97%
(S)-b-Amino-3-fluorobenzenepropanol
Formula: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD17214664
CAS Number: 938462-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR