2,2-Dimethyl-2-fluoroethanol 95%

Supplier: Apollo Scientific
Warning

PC53095-1G
APOSPC53095-1GEA 65.4 EUR
APOSPC53095-1G
2,2-Dimethyl-2-fluoroethanol 95%
2,2-Dimethyl-2-fluoroethanol
Formula: C₄H₉FO
Boiling Pt: 116…118 °C
Density: 0,9681 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD09030962
CAS Number: 3109-99-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR