Zinc(II) trifluoromethanesulphinate 97%

Supplier: Apollo Scientific
Danger

PC53457-1G PC53457-5G
APOSPC53457-1GEA 40.6 EUR
APOSPC53457-1G APOSPC53457-5G
Zinc(II) trifluoromethanesulphinate 97%
Zinc(II) trifluoromethanesulphinate
Formula: C₂F₆O₄S₂Zn
Storage Temperature: Refrigerator
MDL Number: MFCD25978123
CAS Number: 39971-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR