2,6-Difluoro-4-(trifluoromethyl)phenol

Supplier: Apollo Scientific
Danger

PC56034-1G PC56034-250MG
APOSPC56034-1GEA 140 EUR
APOSPC56034-1G APOSPC56034-250MG
2,6-Difluoro-4-(trifluoromethyl)phenol
2,6-Difluoro-4-(trifluoromethyl)phenol
Formula: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD09039179
CAS Number: 276244-25-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR