Potassium (2-acetoxyethyl)trifluoroborate

Supplier: Apollo Scientific
Warning

PC56180-1G PC56180-250MG
APOSPC56180-1GEA 300 EUR
APOSPC56180-1G APOSPC56180-250MG
Potassium (2-acetoxyethyl)trifluoroborate
Potassium (2-acetoxyethyl)trifluoroborate
Formula: C₄H₇BF₃O₂·K
Storage Temperature: Ambient
MDL Number: MFCD22684703
CAS Number: 1408168-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR