2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific
Warning

PC56277-1G PC56277-250MG PC56277-5G
APOSPC56277-1GEA 92.8 EUR
APOSPC56277-1G APOSPC56277-250MG APOSPC56277-5G
2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine 95%
2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine
Formula: C₇H₃F₅INO
Storage Temperature: Ambient
MDL Number: MFCD28789909
CAS Number: 1803729-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR