2-Bromo-6-fluoro-3-(trifluoromethyl)styrene 95%

Supplier: Apollo Scientific
Warning

PC56279-1G PC56279-250MG
APOSPC56279-1GEA 696 EUR
APOSPC56279-1G APOSPC56279-250MG
2-Bromo-6-fluoro-3-(trifluoromethyl)styrene 95%
2-Bromo-6-fluoro-3-(trifluoromethyl)styrene
Formula: C₉H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31613608
CAS Number: 2227272-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR