Methyl 2-bromo-6-fluoronicotinate 95%

Supplier: Apollo Scientific

PC57105-1G PC57105-250MG
APOSPC57105-1GEA 870 EUR
APOSPC57105-1G APOSPC57105-250MG
Methyl 2-bromo-6-fluoronicotinate 95%
Methyl 2-bromo-6-fluoronicotinate
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD14698113
CAS Number: 1214385-74-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR