5-Cyano-2-fluoro-3-methoxyphenylboronic acid pinacol ester 95%

Supplier: Apollo Scientific
Warning

PC5716-100MG PC5716-1G PC5716-250MG
APOSPC5716-100MGEA 219 EUR
APOSPC5716-100MG APOSPC5716-1G APOSPC5716-250MG
5-Cyano-2-fluoro-3-methoxyphenylboronic acid pinacol ester 95%
5-Cyano-2-fluoro-3-methoxyphenylboronic acid pinacol ester
Formula: C₁₄H₁₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD12026087
CAS Number: 1150561-55-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR