3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%

Supplier: Apollo Scientific

PC57808-250MG PC57808-1G PC57808-5G
APOSPC57808-250MGEA 190 EUR
APOSPC57808-250MG APOSPC57808-1G APOSPC57808-5G
3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%
3,5-Dichloro-4-fluoro-2-nitroaniline
Formula: C₆H₃Cl₂FN₂O₂
MDL Number: MFCD30162392
CAS Number: 1698952-59-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR