4-Bromo-3-fluoro-2-methyl-6-nitroaniline ≥95%

Supplier: Apollo Scientific

PC57812-250MG PC57812-1G PC57812-5G
APOSPC57812-250MGEA 116 EUR
APOSPC57812-250MG APOSPC57812-1G APOSPC57812-5G
4-Bromo-3-fluoro-2-methyl-6-nitroaniline ≥95%
4-Bromo-2-fluoro-2-methyl-6-nitroaniline
Formula: C₇H₆BrFN₂O₂
MDL Number: MFCD09152766
CAS Number: 952183-01-6
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR