2,2,3,3,3-Pentafluoropropyl acrylate 97%

Supplier: Apollo Scientific
Danger

Synonyms: 2,2,3,3,3-Pentafluoropropyl prop-2-enoate

PC5785-5G
APOSPC5785-5GEA 116 EUR
APOSPC5785-5G
2,2,3,3,3-Pentafluoropropyl acrylate 97%
2,2,3,3,3-Pentafluoropropyl acrylate
Formula: C₆H₅F₅O₂
Boiling Pt: 50/100 mm °C
Density: 1,32
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039257
CAS Number: 356-86-5
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR