2-Bromo-4,5-difluoroanisole 97%

Supplier: Apollo Scientific
Warning

PC5863-25G PC5863-5G PC5863-10G
APOSPC5863-25GEA 58 EUR
APOSPC5863-25G APOSPC5863-5G APOSPC5863-10G
2-Bromo-4,5-difluoroanisole 97%
2-Bromo-4,5-difluoroanisole
Formula: C₇H₅BrF₂O
Melting Pt: 23…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00070753
CAS Number: 202865-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR