Perfluorobutane-1-sulphonyl fluoride 95%

Supplier: Apollo Scientific
Danger

Synonyms: Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride

PC5887-100G PC5887-250G PC5887-25G
APOSPC5887-100GEA 52.2 EUR
APOSPC5887-100G APOSPC5887-250G APOSPC5887-25G
Perfluorobutane-1-sulphonyl fluoride 95%
Perfluorobutane-1-sulphonyl fluoride
Formula: C₄F₁₀O₂S
Boiling Pt: 64…66 °C
Melting Pt: –110 °C
Density: 1,682 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007422
CAS Number: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR