2-Bromo-5-fluorobenzamide

Supplier: Apollo Scientific
Warning

PC5927-1G
APOSPC5927-1GEA 24.8 EUR
APOSPC5927-1G
2-Bromo-5-fluorobenzamide
2-Bromo-5-fluorobenzamide
Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD03094322
CAS Number: 1006-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR