3-Fluoro-4-(methylsulphonyl)benzonitrile

Supplier: Apollo Scientific
Warning

PC6059-1G PC6059-5G
APOSPC6059-1GEA 132 EUR
APOSPC6059-1G APOSPC6059-5G
3-Fluoro-4-(methylsulphonyl)benzonitrile
3-Fluoro-4-(methylsulphonyl)benzonitrile
Formula: C₈H₆FNO₂S
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD12922838
CAS Number: 185946-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR