4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Cyano-2-(methylsulphonyl)benzotrifluoride, 4-Cyano-2-(trifluoromethyl)phenyl methyl sulphone

PC6219-1G PC6219-5G
APOSPC6219-1GEA 415 EUR
APOSPC6219-1G APOSPC6219-5G
4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile 98%
4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO₂S
Melting Pt: 171…172 °C
Storage Temperature: Ambient
MDL Number: MFCD12196930
CAS Number: 904311-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR