1H,1H-Heneicosafluoro-1-undecanol 95+%

Supplier: Apollo Scientific
Warning

PC6222C-1G
APOSPC6222C-1GEA 27.7 EUR
APOSPC6222C-1G
1H,1H-Heneicosafluoro-1-undecanol 95+%
1H,1H-Heneicosafluoro-1-undecanol
Formula: C₁₁H₃F₂₁O
Boiling Pt: 143 °C/80mmHg
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00153269
CAS Number: 307-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR