Potassium fluoride dihydrate 98%

Supplier: Apollo Scientific
Danger

PC6267R-250G PC6267R-50G
APOSPC6267R-250GEA 32.2 EUR
APOSPC6267R-250G APOSPC6267R-50G
Potassium fluoride dihydrate 98%
Potassium fluoride dihydrate
Formula: FK·2H₂O
Boiling Pt: 156 °C
Melting Pt: 41 °C
Density: 2,454 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149929
CAS Number: 13455-21-5
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR