Tetrafluorohydroquinone 95%

Supplier: Apollo Scientific
Danger

PC6790-1G PC6790-25G PC6790-5G
APOSPC6790-1GEA 43.5 EUR
APOSPC6790-1G APOSPC6790-25G APOSPC6790-5G
Tetrafluorohydroquinone 95%
Tetrafluorohydroquinone
Formula: C₆H₂F₄O₂
Melting Pt: 168…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00002340
CAS Number: 771-63-1
EINECS: 212-237-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR