Tetrafluoroisophthalonitrile 98%

Supplier: Apollo Scientific
Danger

PC6797-1G PC6797-25G PC6797-5G
APOSPC6797-1GEA 21.9 EUR
APOSPC6797-1G APOSPC6797-25G APOSPC6797-5G
Tetrafluoroisophthalonitrile 98%
Tetrafluoroisophthalonitrile
Formula: C₈F₄N₂
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00013290
CAS Number: 2377-81-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR