Lithium bis(trifluoromethylsulphonyl)imide 99%

Supplier: Apollo Scientific
Danger

PC6834-50G PC6834-5G PC6834-25G PC6834-100G
APOSPC6834-50GEA 58 EUR
APOSPC6834-50G APOSPC6834-5G APOSPC6834-25G APOSPC6834-100G
Lithium bis(trifluoromethylsulphonyl)imide 99%
Lithium bis(trifluoromethylsulfonyl)imide
Formula: C₂F₆LiNO₄S₂
Melting Pt: 234…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00210017
CAS Number: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR