2,3-Difluoro-4-methylphenacyl bromide

Supplier: Apollo Scientific
Danger

PC6884-1G PC6884-250MG
APOSPC6884-1GEA 1050 EUR
APOSPC6884-1G APOSPC6884-250MG
2,3-Difluoro-4-methylphenacyl bromide
2,3-Difluoro-4-methylphenacyl bromide
Formula: C₉H₇BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD14581695
CAS Number: 1221684-44-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR