Triethylsilyl trifluoromethanesulphonate 99%

Supplier: Apollo Scientific
Danger

PC7114-100G PC7114-25G PC7114-5G
APOSPC7114-100GEA 60.9 EUR
APOSPC7114-100G APOSPC7114-25G APOSPC7114-5G
Triethylsilyl trifluoromethanesulphonate 99%
Triethylsilyl trifluoromethanesulphonate

Lewis acid. Powerful silylating reagent .

Formula: C₇H₁₅F₃O₃SSi
Boiling Pt: 207 °C
Density: 1,169 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR