4-Carbamoyl-3-fluorobenzeneboronic acid 96%

Supplier: Apollo Scientific
Warning

PC7136-1G
APOSPC7136-1GEA 156 EUR
APOSPC7136-1G
4-Carbamoyl-3-fluorobenzeneboronic acid 96%
4-Carbamoyl-3-fluorobenzeneboronic acid
Formula: C₇H₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD09258667
CAS Number: 874288-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR