3-Cyano-1-propylboronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile

PC7178-25G PC7178-5G PC7178-1G
APOSPC7178-25GEA 1200 EUR
APOSPC7178-25G APOSPC7178-5G APOSPC7178-1G
3-Cyano-1-propylboronic acid pinacol ester
3-Cyano-1-propylboronic acid pinacol ester
Formula: C₁₀H₁₈BNO₂
MW: 195.07 g/mol
Boiling Pt: 85 °C/0.5mmHg
Storage Temperature: Freezer
MDL Number: MFCD09953480
CAS Number: 238088-16-9
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR