α,α,α-Trifluoroacetophenone 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

PC7200-100G PC7200-25G PC7200-5G
APOSPC7200-100GEA 122 EUR
APOSPC7200-100G APOSPC7200-25G APOSPC7200-5G
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Formula: C₈H₅F₃O
Boiling Pt: 165…166 °C
Density: 1,24 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR